missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ GSK 2239633A
Description
GSK 2239633A is a potent CCR4 antagonist (pIC50 = 7.83 in a 35S-GTPγS competition assay). Inhibits thymus activation-regulated chemokine (TARC) -induced F-actin polymerization in human whole blood.
Specifications
Specifications
| Chemical Name or Material | N-[[3-[[3-[[(5-Chloro-2-thienyl)sulfonyl]amino]-4-methoxy-1H-indazol-1-yl]methyl]phenyl]methyl]-2-hydroxy-2-methylpropanamide |
| CAS | 1240516-71-5 |
| Quantity | 10 mg |
| Molecular Formula | C24H25ClN4O5S2 |
| Purity | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?