missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ GSK 2194069
GREENER_CHOICE

Product Code. 18716858
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18716858 25 mg 25mg
18780428 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18716858 Supplier Tocris Bioscience™ Supplier No. 5303/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent human fatty acid synthase (hFASN) inhibitor

GSK 2194069 is a potent human fatty acid synthase (hFASN) inhibitor (IC50 = 7.7 nM); inhibits hFAS β-ketoacyl reductase activity. Inhibits lipid synthesis and attenuates proliferation of A549 non-small-cell lung cancer cells (EC50 = 15 nM) in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-[4-(5-Benzofuranyl)phenyl]-5-[[(3S)-1-(cyclopropylcarbonyl)-3-pyrrolidinyl]methyl]-2,4-dihydro-3H-1,2,4-triazol-3-one
CAS 1332331-08-4
Assay Percent Range >99%
Quantity 25 mg
Molecular Formula C25H24N4O3
Formula Weight Observed MW: 428.48
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.