missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ GSK 2193874

Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
GSK 2193874 is a potent and selective TRPV4 antagonist (IC50 values are 2 and 40 nM for rat and human receptors, respectively); inhibits Ca2+ influx through TRPV4 channels. Prevents and reverses pulmonary edema after myocardial infarction in vivo models. Selective over a panel of ∽200 human receptors, channels and enzymes. Orally active.
Specifications
Specifications
| Chemical Name or Material | 3-([1,4'-Bipiperidin]-1'-ylmethyl)-7-bromo-N-(1-phenylcyclopropyl)-2-[3-(trifluoromethyl)phenyl]-4-quinolinecarboxamide |
| CAS | 1336960-13-4 |
| Assay Percent Range | >98% |
| Quantity | 5 mg |
| Molecular Formula | C37H38BrF3N4O |
| Formula Weight | Observed MW: 691.62 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction