missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ GSK 2033
Description
GSK 2033 is a potent LXR antagonist (pIC50 = 7.5). Enhances T-cell proliferation and blocks T 0901317 -antiproliferative activity on T-cells. Cell permeable.
Specifications
Specifications
| Chemical Name or Material | 2,4,6-Trimethyl-N-[[3′-(methylsulfonyl)[1,1′-biphenyl]-4-yl]methyl]-N-[[5-(trifluoromethyl)-2-furanyl]methyl]benzenesulfonamide |
| CAS | 1221277-90-2 |
| Quantity | 10 mg |
| Target | LXR-like Receptor Antagonists |
| Molecular Formula | C29H28F3NO5S |
| Formula Weight | 591.66 |
| Purity | >99% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?