missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ GSK 126
GREENER_CHOICE

Product Code. 18787606
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18787606 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18787606 Supplier Tocris Bioscience™ Supplier No. 6790/5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Very high affinity and selective EZH2 inhibitor

GSK 126 is a very high affinity and selective EZH2 inhibitor (Ki = 0.5 - 3 nM). Exhibits >150-fold selectivity for EZH2 over EZH1 and >1000-fold selectivity over 20 other human methyltransferases. Inhibits proliferation of B-cell lymphoma cell lines in vitro. Also inhibits H3K27Me3 and tumor growth in mice bearing BCL xenografts. Targeting p53 and histone methyltransferases restores exhausted CD8+ T cells in HCV infection. Promotes the transition of ESCs to feeder-free extended pluripotent stem (EPS) cells in combination with LCDM cocktail .
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[(1,2-Dihydro-4,6-dimethyl-2-oxo-3-pyridinyl)methyl]-3-methyl-1-[(1S)-1-methylpropyl]-6-[6-(1-piperazinyl)-3-pyridinyl]-1H-indole-4-carboxamide
CAS 1346574-57-9
Quantity 5 mg
Molecular Formula C31H38N6O2
Formula Weight 526.67
Product Type Inhibitor
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.