missing translation for 'onlineSavingsMsg'
Learn More

GSK-1070916, MedChemExpress

Product Code. p-200101683
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16494077

Brand: Medchem Express HY70044

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GSK-1070916 is a potent and selective ATP-competitive inhibitor of aurora B and aurora C with Kis of 0.38 and 1.5 nM, respectively, and is >250- fold selective over Aurora A.

GSK-1070916 is a potent and selective ATP-competitive inhibitor of aurora B and aurora C with Kis of 0.38 and 1.5 nM, respectively, and is >250- fold selective over Aurora A.

Specifications

Chemical Name or Material GSK-1070916
Physical Form Solid
CAS 942918-07-2
Color Orange
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C30H33N7O
For Use With (Application) Cancer-Kinase/protease
Synonym GSK-1070916A
Solubility Information DMSO : 16.67 mg/mL (32.84 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(NC1=CC=C(C2=NN(C=C2C3=C4C(NC(C5=CC=CC(CN(C)C)=C5)=C4)=NC=C3)CC)C=C1)N(C)C
Molecular Weight (g/mol) 507.63
Formula Weight 507.63
Percent Purity 98.06%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.