missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ GSK 0660
GREENER_CHOICE

Product Code. 18094182
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18094182 50 mg 50mg
18033972 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18094182 Supplier Tocris Bioscience™ Supplier No. 3433/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective PPARδ antagonist

Selective PPARδ antagonist (IC50 values are 0.155, > 10 and ≥ 10 μM at PPARδ, PPARα and PPARγ respectively). Exhibits inverse agonist effects when administered by itself.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1014691-61-2
Molecular Formula C19H18N2O5S2

Specifications

Quantity 50 mg
Formula Weight 418.49
Purity >99%
Molecular Weight (g/mol) 418.48
Chemical Name or Material GSK 0660
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.