missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ GSK 0660

Description
Selective PPARδ antagonist (IC50 values are 0.155, > 10 and ≥ 10 μM at PPARδ, PPARα and PPARγ respectively). Exhibits inverse agonist effects when administered by itself.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 418.49 |
| Purity | >99% |
| Molecular Weight (g/mol) | 418.48 |
| Chemical Name or Material | GSK 0660 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?