Learn More
GSG2 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55454-1-AP-20UL
This item is not returnable.
View return policy
Description
The GSG2 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human GSG2. This antibody recognizes human antigen. The GSG2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| GSG2 | |
| 400 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| GSG2, H haspin, HASPIN | |
| 20 μL | |
| 83903 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |