Learn More
Invitrogen™ Human GSG2 (Haspin), GST Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Catalytic
Specifications
Specifications
| Accession Number | Q8TF76 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50 mM tris with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5 |
| Gene ID (Entrez) | 83903 |
| Molecular Weight (g/mol) | 64.8 kDa |
| Name | Human GSG2 (Haspin), GST Tag |
| Quantity | 10 μg |
| Gene Alias | germ cell associated 2 (haspin); germ cell associated 2, haspin; germ cell-specific gene 2 protein; GSG2; haploid germ cell-specific nuclear protein kinase; HASPIN; H-haspin; Serine/threonine-protein kinase haspin |
| Common Name | GSG2 |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.