missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSG2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35922-100ul
This item is not returnable.
View return policy
Description
GSG2 Polyclonal antibody specifically detects GSG2 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| GSG2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, ELISA | |
| EC 2.7.11.1, germ cell associated 2 (haspin), Germ cell-specific gene 2 protein, Haploid germ cell-specific nuclear protein kinase, HASPIN, H-haspin, serine/threonine-protein kinase haspin | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 290-470 of human GSG2 (NP_114171.2).,, Sequence:, GKRRATGQDSCQERGLQEAVRREHQEASVPKGRIVPRGIDRLERTRSSRKSKHQEATETSLLHSHRFKKGQKLGKDSFPTQDLTPLQNVCFWTKTRASFSFHKKKIVTDVSEVCSIYTTATSLSGSLLSECSNRPVMNRTSGAPSSWHSSSMYLLSPLNTLSISNKKASDAEKVYGECSQK | |
| 100 μL | |
| Cell Cycle and Replication | |
| 83903 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction