missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSG2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-26626
This item is not returnable.
View return policy
Description
GSG2 Polyclonal specifically detects GSG2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| GSG2 | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| EC 2.7.11.1, germ cell associated 2 (haspin), Germ cell-specific gene 2 protein, Haploid germ cell-specific nuclear protein kinase, HASPIN, H-haspin, serine/threonine-protein kinase haspin | |
| Rabbit | |
| 88 kDa | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 83903 | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| Western Blot 1:2000-1:10000, Immunohistochemistry 1:100-1:500, Immunoprecipitation 5-10 ug/mg lysate, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Q8TF76 | |
| GSG2 | |
| The immunogen recognized by this antibody maps to a region between residue 300 and 350 of human serine/threonine-protein kinase Haspin (germ cell-specific gene 2) using the numbering given in entry NP_114171.2 (GeneID 83903). | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction