missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSE1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA557628
This item is not returnable.
View return policy
Description
Immunogen sequence: LDTSSEKLEF LQLFGLTTQQ QKEELVAQKR RKRRRMLRER SPSPPTIQSK RQTPSPRLAL STRYSPDEMN NSPNFEEKKK FLTIFNLTH Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 96%.
This gene encodes a proline-rich protein with coiled coil domains that may be a subunit of a BRAF35-HDAC (BHC) histone deacetylase complex. This gene may function as an oncogene in breast cancer and enhanced expression of the encoded protein has been observed in breast cancer patients.
Specifications
| GSE1 | |
| Polyclonal | |
| Unconjugated | |
| GSE1 | |
| 2210013I18Rik; CRHSP24; CTC-786C10.1; fj87f12; genetic suppressor element 1; GSE1; Gse1 coiled-coil protein; Gse1 coiled-coil protein homolog; Kiaa0182; mKIAA0182; RGD1562686; si:dkey-265o13.4; wu:fj87f12 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23199 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q14687 | |
| GSE1 | |
| Recombinant protein corresponding to Human GSE1 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction