missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSE1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-79573
This item is not returnable.
View return policy
Description
GSE1 Polyclonal specifically detects GSE1 in Human samples. It is validated for Western Blot.
Specifications
| GSE1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| KIAA0182 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 23199 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| NP_055430 | |
| GSE1 | |
| Synthetic peptide directed towards the N terminal of human KIAA0182The immunogen for this antibody is KIAA0182. Peptide sequence EEPRGSSLSSESSPVSSPATNHSSPASTPKRVPMGPIIVPPGGHSVPSTP. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Xenopus: 100%; Bovine: 100%; Chicken: 100%; Canine: 100%; Guinea pig: 100%; Equine: 100%; Pig: 100%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction