missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ GSDMDC1 Antibody Pair

Product Code. 18425089 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18425089 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18425089 Supplier Novus Biologicals™ Supplier No. H00079792AP21

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Assay Range 0.1 ng/mL to 100 ng/mL
Conjugate Unconjugated
Product Type Sandwich ELISA
For Use With (Application) Sandwich ELISA
Gene ID (Entrez) 79792
Gene Symbol GSDMD
Quantity 1 Set
Research Discipline Cell Biology
Storage Requirements Aliquot and store at -20°C to +80°C. Avoid freeze/thaw cycles.
Synonym DF5L, DFNA5L, FLJ12150, gasdermin D, gasdermin domain containing 1, Gasdermin domain-containing protein 1, gasdermin-D, GSDMD N, GSDMDC1, GSDMD-N
Target Human
Buffer 1 x PBS, pH 7.4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.