missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GSC Monoclonal Antibody (4C5D10)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA548806
This item is not returnable.
View return policy
Description
GSC Monoclonal Antibody for Western Blot, IHC, Flow, ELISA
This gene encodes a member of the bicoid subfamily of the paired (PRD) homeobox family of proteins. The encoded protein acts as a transcription factor and may be autoregulatory. A similar protein in mice plays a role in craniofacial and rib cage development during embryogenesis.
Specifications
| GSC | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| P56915 | |
| GSC | |
| Purified recombinant fragment of human GSC (AA: 191-257) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunohistochemistry, Western Blot | |
| 4C5D10 | |
| Unconjugated | |
| GSC | |
| GCS; goosecoid; goosecoid homeobox; goosecoid protein; GSC; Homeobox protein goosecoid; SAMS | |
| Mouse | |
| Protein G | |
| RUO | |
| 145258 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction