missing translation for 'onlineSavingsMsg'
Learn More

GSAP Antibody, DyLight 594, Novus Biologicals™

Product Code. 18433547 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18433547 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18433547 Supplier Novus Biologicals Supplier No. NBP178376DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GSAP Polyclonal antibody specifically detects GSAP in Human samples. It is validated for Western Blot, Simple Western
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSAP
Applications Western Blot, Simple Western
Classification Polyclonal
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias pigeon homolog (Drosophila)
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human GSAP protein (between residues 100-150) [UniProt A4D1B5]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 54103
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.