missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSAP Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-78376CL1
This item is not returnable.
View return policy
Description
GSAP Polyclonal antibody specifically detects GSAP in Human samples. It is validated for Western Blot,Simple Western
Specifications
| GSAP | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Simple Western | |
| CoraFluor 1 | |
| pigeon homolog (Drosophila) | |
| A synthetic peptide made to an internal portion of the human GSAP protein (between residues 100-150) [UniProt A4D1B5] | |
| 0.1 mL | |
| Alzheimers Research, Neurodegeneration, Neuroscience | |
| 54103 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction