missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSAP Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81894
This item is not returnable.
View return policy
Description
GSAP Polyclonal specifically detects GSAP in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| GSAP | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| PION | |
| GSAP antibody was raised against a 19 amino acid synthetic peptide near the carboxy terminus of human GSAP. The immunogen is located within amino acids 770 - 820 of GSAP. Amino Acid Squence: RNHVTRLLQNYKKQPRNSM | |
| 0.1 mg | |
| Alzheimers Research, Neurodegeneration, Neuroscience | |
| 54103 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 0.25 ug/ml, ELISA, Immunohistochemistry 5 ug/mL, Immunocytochemistry/Immunofluorescence 20 ug/mL, Immunohistochemistry-Paraffin | |
| pigeon homolog (Drosophila) | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Multiple isoforms of GSAP are known to exist. GSAP antibody is predicted to not cross-react with other F-box protein family members. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction