missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies GSA 10
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
- GSA 10 is an agonist of Smoothened (SMO), a cell surface receptor and mediator of the hedgehog signaling pathway
- It is a quinolinecarboxamide derivative that binds in a distinct binding pocket from cyclopamine
- GSA10 is active at an EC50 of 1.2 μM in an alkaline phosphatase-based mouse mesenchymal cell line (C3H10T1/2) differentiation assay (Gorojankina et al.)
- Promotes differentiation of multipotent mesenchymal progenitor cells into osteoblasts (Gorojankina et al.)
Specifications
Specifications
| CAS | 300833-95-8 |
| Quantity | 50 mg |
| Molecular Formula | C26H30N2O5 |
| Purity | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction