missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies GSA 10

Product Code. 17140182
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 17140182

Brand: STEMCELL Technologies 73172

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Hedgehog pathway activator; Activates Smoothened (SMO)

  • GSA 10 is an agonist of Smoothened (SMO), a cell surface receptor and mediator of the hedgehog signaling pathway
  • It is a quinolinecarboxamide derivative that binds in a distinct binding pocket from cyclopamine
  • GSA10 is active at an EC50 of 1.2 μM in an alkaline phosphatase-based mouse mesenchymal cell line (C3H10T1/2) differentiation assay (Gorojankina et al.)
  • Promotes differentiation of multipotent mesenchymal progenitor cells into osteoblasts (Gorojankina et al.)

Specifications

CAS 300833-95-8
Quantity 10 mg
Molecular Formula C26H30N2O5
Purity ≥95%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.