missing translation for 'onlineSavingsMsg'
Learn More

GS967, MedChemExpress

Product Code. p-200099528
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16481467 50 mg 50mg
16475258 10 mg 10mg
16437977 5 mg 5mg
16431046 200 mg 200mg
16425926 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16481467 Supplier Medchem Express Supplier No. HY12593

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GS967 (GS-458967) is a potent, and selective inhibitor of cardiac late sodium current (late INa ) with IC50 values of 0.13 and 0.21 μM for ventricular myocytes and isolated hearts, respectively.

GS967 (GS-458967) is a potent, and selective inhibitor of cardiac late sodium current (late INa ) with IC50 values of 0.13 and 0.21 μM for ventricular myocytes and isolated hearts, respectively.

Specifications

Chemical Name or Material GS967
CAS 1262618-39-2
Color White
Purity Grade Notes Research
Molecular Formula C14H7F6N3O
Quantity 50 mg
Solubility Information DMSO : 50 mg/mL (144.00 mM; Need ultrasonic)
SMILES FC(C1=NN=C2C=CC(C3=CC=C(OC(F)(F)F)C=C3)=CN21)(F)F
Molecular Weight (g/mol) 347.22
Formula Weight 347.22
Percent Purity 99.95%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.