missing translation for 'onlineSavingsMsg'
Learn More

GS28 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16835604 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16835604 20 μL 20µL
16825604 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16835604 Supplier Proteintech Supplier No. 161061AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This gene encodes a trafficking membrane protein which transports proteins among the endoplasmic reticulum and the Golgi and between Golgi compartments. This protein is considered an essential component of the Golgi SNAP receptor (SNARE) complex. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.

Specifications

Antigen GS28
Applications Immunohistochemistry (Paraffin), Flow Cytometry, Immunofluorescence, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GOSR1
Gene Accession No. O88630, O95249, Q62931
Gene Alias 28 kDa cis Golgi SNARE p28, 28 kDa Golgi SNARE protein, GOLIM2, GOS 28, GOS28, GOS28/P28, GOSR1, GS28, P28
Gene Symbols Gosr1
Host Species Rabbit
Immunogen GS28 Fusion Protein Ag9021
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53334, 94189, 9527
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.