missing translation for 'onlineSavingsMsg'
Learn More

GS-444217, MedChemExpress

Product Code. 16484466
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16484466 100 mg 100mg
16475917 5 mg 5mg
16469736 50 mg 50mg
16453188 10 mg 10mg
16420709 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16484466 Supplier Medchem Express Supplier No. HY100844
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress GS-444217 is a potent, orally available and selective ATP-competitive inhibitor of apoptosis signal-regulating kinase 1 (ASK1) with an IC50 of 2.87 nM.

GS-444217 is a potent, orally available and selective ATP-competitive inhibitor of apoptosis signal-regulating kinase 1 (ASK1) with an IC50 of 2.87 nM.

Specifications

Chemical Name or Material GS-444217
CAS 1262041-49-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C23H21N7O
Quantity 100 mg
Solubility Information DMSO : 15.5 mg/mL (37.67 mM; Need ultrasonic and warming)
SMILES O=C(C1=NC=CC(N2C=C(C3CC3)N=C2)=C1)NC4=CC=CC(C5=NN=CN5C6CC6)=C4
Molecular Weight (g/mol) 411.46
Formula Weight 411.46
Percent Purity 99.43%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.