missing translation for 'onlineSavingsMsg'
Learn More

GS-444217, MedChemExpress

Product Code. 16453188
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16453188

Brand: Medchem Express HY100844

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GS-444217 is a potent, orally available and selective ATP-competitive inhibitor of apoptosis signal-regulating kinase 1 (ASK1) with an IC50 of 2.87 nM.

GS-444217 is a potent, orally available and selective ATP-competitive inhibitor of apoptosis signal-regulating kinase 1 (ASK1) with an IC50 of 2.87 nM.

Specifications

Chemical Name or Material GS-444217
CAS 1262041-49-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C23H21N7O
Quantity 10 mg
Solubility Information DMSO : 15.5 mg/mL (37.67 mM; Need ultrasonic and warming)
SMILES O=C(C1=NC=CC(N2C=C(C3CC3)N=C2)=C1)NC4=CC=CC(C5=NN=CN5C6CC6)=C4
Molecular Weight (g/mol) 411.46
Formula Weight 411.46
Percent Purity 99.43%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.