missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ GS 39783

Product Code. 30001313
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001313 10 mg 10mg
18034722 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001313 Supplier R&D Systems™ Supplier No. 2001/10
  • ONLINE EXCLUSIVE PRICE
    £171.71
    £153.30 / 10mg
    Save £18.41
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Positive allosteric modulator of GABAB receptors

Positive allosteric modulator of GABAB receptor function. Potentiates the effects of GABA on [35S]GTPγS binding at recombinant and native GABAB receptors (EC50 values are 2.1 and 3.1μM respectively). Decreases cocaine self-administration, blocks the rewarding properties of nicotine and produces anxiolytic-like activity without the side effects associated with baclofen or benzodiazepines.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39069-52-8
Molecular Formula C15H23N5O2S

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 337.44
Purity >98%
Molecular Weight (g/mol) 337.442
Chemical Name or Material GS 39783
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.