missing translation for 'onlineSavingsMsg'
Learn More

Grubbs Catalyst, 2nd Generation, TRC

Product Code. 30393953
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393953 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30393953 Supplier Toronto Research Chemicals Supplier No. TRCG7890001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Grubbs Catalyst, 2nd Generation
CAS 246047-72-3
Molecular Formula C28 H32 Cl2 N2 Ru . C18 H33 P
Quantity 1 g
Synonym Ruthenium, [1,3-bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene]dichloro(phenylmethylene)(tricyclohexylphosphine)-, (SP-5-41)- (9CI, ACI), (SP-5-41)-[1,3-Bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene]dichloro(phenylmethylene)(tricyclohexylphosphine)ruthenium (ACI), (1,3-Dimesitylimidazolidin-2-ylidene)(tricyclohexylphosphine)benzylidene ruthenium dichloride, [1,3-Bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene]dichloro(phenylmethylene)(tricyclohexylphosphine)ruthenium, Benzylidene(1,3-dimesityl-4-imidazolidin-2-ylidene)(tricyclohexylphosphine)ruthenium dichloride, Benzylidene(1,3-dimesitylimidazolidin-2-ylidene)(tricyclohexylphosphine)ruthenium dichloride, Benzylidene[1,3-bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene]dichloro(tricyclohexylphosphine)ruthenium, Benzylidenedichloro(1,3-dimesitylimidazolidin-2-ylidene)(tricyclohexylphosphine )ruthenium, C 848, C 848 (catalyst), Dichloro(1,3-dimesityl-4,5-dihydroimidazol-2-ylidene)(phenylmethylene)(tricyclohexylphosphine)ruthenium, Generation II Grubbs' catalyst, Grubbs 2nd catalyst, Grubbs 2nd generation catalyst, Grubbs Catalyst 2nd. generation, Grubbs II catalyst, Grubbs second generation catalyst, Grubbs' 2nd generation catalyst, Grubbs' Ru-dihydroimidazolylidene catalyst, Grubbs-Herrmann catalyst, Grubbs-Nolan catalyst, M 204, Second generation Grubbs catalyst, Secondary generation Grubbs catalyst, Tricyclohexylphosphine[1,3-bis(2,4,6-trimethylphenyl)-4,5-dihydroimidazol-2-ylidene](benzylidene)ruthenium(IV) dichloride, Tricyclohexylphosphine[1,3-bis(2,4,6-trimethylphenyl)-4,5-dihydroimidazol-2-ylidene]benzylidene ruthenium dichloride, Tricyclohexylphosphino[1,3-bis(2,4,6-trimethylphenyl)-4,5-dihydroimidazol-2-ylidene]benzylidene ruthenium dichloride
InChI Formula InChI=1S/C21H26N2.C18H33P.C7H6.2ClH.Ru/c1-14-9-16(3)20(17(4)10-14)22-7-8-23(13-22)21-18(5)11-15(2)12-19(21)6;1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;1-7-5-3-2-4-6-7;;;/h9-12H,7-8H2,1-6H3;16-18H,1-15H2;1-6H;2*1H;/q;;;;;+2/p-2
SMILES Cc1cc(C)c(N2CCN(C2=[Ru](=Cc3ccccc3)(Cl)Cl)c4c(C)cc(C)cc4C)c(C)c1.C5CCC(CC5)P(C6CCCCC6)C7CCCCC7
IUPAC Name benzylidene-[1,3-bis(2,4,6-trimethylphenyl)imidazolidin-2-ylidene]-dichlororuthenium;tricyclohexylphosphane
Molecular Weight (g/mol) 848.97
Formula Weight 848.3306
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.