missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Grubbs Catalyst, 2nd Generation, TRC
Specifications
Specifications
| Chemical Name or Material | Grubbs Catalyst, 2nd Generation |
| CAS | 246047-72-3 |
| Molecular Formula | C28 H32 Cl2 N2 Ru . C18 H33 P |
| Quantity | 250 mg |
| Synonym | Ruthenium, [1,3-bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene]dichloro(phenylmethylene)(tricyclohexylphosphine)-, (SP-5-41)- (9CI, ACI), (SP-5-41)-[1,3-Bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene]dichloro(phenylmethylene)(tricyclohexylphosphine)ruthenium (ACI), (1,3-Dimesitylimidazolidin-2-ylidene)(tricyclohexylphosphine)benzylidene ruthenium dichloride, [1,3-Bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene]dichloro(phenylmethylene)(tricyclohexylphosphine)ruthenium, Benzylidene(1,3-dimesityl-4-imidazolidin-2-ylidene)(tricyclohexylphosphine)ruthenium dichloride, Benzylidene(1,3-dimesitylimidazolidin-2-ylidene)(tricyclohexylphosphine)ruthenium dichloride, Benzylidene[1,3-bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene]dichloro(tricyclohexylphosphine)ruthenium, Benzylidenedichloro(1,3-dimesitylimidazolidin-2-ylidene)(tricyclohexylphosphine )ruthenium, C 848, C 848 (catalyst), Dichloro(1,3-dimesityl-4,5-dihydroimidazol-2-ylidene)(phenylmethylene)(tricyclohexylphosphine)ruthenium, Generation II Grubbs' catalyst, Grubbs 2nd catalyst, Grubbs 2nd generation catalyst, Grubbs Catalyst 2nd. generation, Grubbs II catalyst, Grubbs second generation catalyst, Grubbs' 2nd generation catalyst, Grubbs' Ru-dihydroimidazolylidene catalyst, Grubbs-Herrmann catalyst, Grubbs-Nolan catalyst, M 204, Second generation Grubbs catalyst, Secondary generation Grubbs catalyst, Tricyclohexylphosphine[1,3-bis(2,4,6-trimethylphenyl)-4,5-dihydroimidazol-2-ylidene](benzylidene)ruthenium(IV) dichloride, Tricyclohexylphosphine[1,3-bis(2,4,6-trimethylphenyl)-4,5-dihydroimidazol-2-ylidene]benzylidene ruthenium dichloride, Tricyclohexylphosphino[1,3-bis(2,4,6-trimethylphenyl)-4,5-dihydroimidazol-2-ylidene]benzylidene ruthenium dichloride |
| InChI Formula | InChI=1S/C21H26N2.C18H33P.C7H6.2ClH.Ru/c1-14-9-16(3)20(17(4)10-14)22-7-8-23(13-22)21-18(5)11-15(2)12-19(21)6;1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;1-7-5-3-2-4-6-7;;;/h9-12H,7-8H2,1-6H3;16-18H,1-15H2;1-6H;2*1H;/q;;;;;+2/p-2 |
| SMILES | Cc1cc(C)c(N2CCN(C2=[Ru](=Cc3ccccc3)(Cl)Cl)c4c(C)cc(C)cc4C)c(C)c1.C5CCC(CC5)P(C6CCCCC6)C7CCCCC7 |
| IUPAC Name | benzylidene-[1,3-bis(2,4,6-trimethylphenyl)imidazolidin-2-ylidene]-dichlororuthenium;tricyclohexylphosphane |
| Molecular Weight (g/mol) | 848.97 |
| Formula Weight | 848.3306 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?