missing translation for 'onlineSavingsMsg'
Learn More

Grubbs Catalyst, 1st Generation, TRC

Product Code. 30392302
Change view
Click to view available options
Quantity:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30392302 1 g
30407907 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30392302 Supplier Toronto Research Chemicals Supplier No. TRCG7890051G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 172222-30-9
Molecular Formula C43H72Cl2P2Ru
Molecular Weight (g/mol) 822.96
Synonym Grubbs Catalyst, 1st Generation, Dichloro(phenylmethylene)bis(tricyclohexylphosphine)ruthenium, Grubbs' ruthenium catalyst, Ruthenium(II)dichloro(phenylmethylene)bis(tricyclohexylphosphine), Benzylidenedichlorobis(tricyclohexylphosphine)ruthenium
SMILES [Cl-][Ru+4](=[CH-2]c1ccccc1)([Cl-])([P](C2CCCCC2)(C3CCCCC3)C4CCCCC4)[P](C5CCCCC5)(C6CCCCC6)C7CCCCC7

Specifications

Quantity 1 g
InChI Formula InChI=1S/2C18H33P.C7H6.2ClH.Ru/c2*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;1-7-5-3-2-4-6-7;;;/h2*16-18H,1-15H2;1-6H;2*1H;/q;;-2;;;+4/p-2
Formula Weight 822.353
Chemical Name or Material Benzylidenebis(tricyclohexylphosphine)dichlororuthenium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.