missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRTP1 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-76900CL1
This item is not returnable.
View return policy
Description
GRTP1 Polyclonal antibody specifically detects GRTP1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| GRTP1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 79774 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| FLJ22474, growth hormone regulated TBC protein 1, growth hormone-regulated TBC protein 1, MGC138328, TBC1 domain family member 6, TBC1 domain family, member 6, TBC1D6MGC138330 | |
| Antibody was raised against a 18 amino acid synthetic peptide from near the amino terminus of human GRTP1. The immunogen is located within the first 50 amino acids of GRTP1. Amino Acid Squence: IDPYGFERPEDFDDAAYE | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction