missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRTP1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76376
This item is not returnable.
View return policy
Description
GRTP1 Polyclonal specifically detects GRTP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.
Specifications
| GRTP1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| FLJ22474, growth hormone regulated TBC protein 1, growth hormone-regulated TBC protein 1, MGC138328, TBC1 domain family member 6, TBC1 domain family, member 6, TBC1D6MGC138330 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 79774 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, ELISA | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000 | |
| EAX09206 | |
| GRTP1 | |
| Antibody was raised against a 16 amino acid synthetic peptide from near the center of human GRTP1. The immunogen is located within amino acids 80 - 130 of GRTP1. Amino Acid Squence: LQGERNPRLEDAIRTD | |
| 0.1 mg | |
| Primary | |
| At least three isoforms of GRTP1 are known to exist; this antibody should recognize all isoforms. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering