missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRSF1 Antibody (JE65-34), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32410
This item is not returnable.
View return policy
Description
GRSF1 Monoclonal antibody specifically detects GRSF1 in Human samples. It is validated for Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| GRSF1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500, Immunocytochemistry/Immunofluorescence 1:50 | |
| FLJ13125, G-rich RNA sequence binding factor 1, G-rich sequence factor 1, GRSF-1 | |
| Synthetic peptide within human GRSF1 aa 350-400/480. (Uniprot: Q12849) | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| JE65-34 | |
| Unconjugated | |
| 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 2926 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction