missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GRPEL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553747
This item is not returnable.
View return policy
Description
Immunogen sequence: EPEDQKLTLE KVFRGLLLLE AKLKSVFAKH GLEKLTPIGD KYDPHEHELI CHVPAGVGVQ PGTVALVRQD GYKLHGRTIR LARVEVAVES Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 90%.
GRPEL2 (GrpE Like 2, Mitochondrial) is a Protein Coding gene. Among its related pathways are Mitochondrial protein import and Metabolism of proteins. Gene Ontology (GO) annotations related to this gene include protein homodimerization activity and chaperone binding. An important paralog of this gene is GRPEL1. Essential component of the PAM complex, a complex required for the translocation of transit peptide-containing proteins from the inner membrane into the mitochondrial matrix in an ATP-dependent manner. Seems to control the nucleotide-dependent binding of mitochondrial HSP70 to substrate proteins. Stimulates ATPase activity of mt-HSP70. May also serve to modulate the interconversion of oligomeric (inactive) and monomeric (active) forms of mt-HSP70.
Specifications
| GRPEL2 | |
| Polyclonal | |
| Unconjugated | |
| GRPEL2 | |
| actin filament-associated protein 1-like 1; Afap1l1; AFAP1-like protein 1; GrpE like 2, mitochondrial; grpE protein homolog 2, mitochondrial; Grpel2; GrpE-like 2, mitochondrial; Mt-GrpE#2; Mt-grpel2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 134266 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TAA5 | |
| GRPEL2 | |
| Recombinant protein corresponding to Human GRPEL2. Recombinant protein control fragment (Product #RP-91014). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction