missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GRPEL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542859
This item is not returnable.
View return policy
Description
Peptide sequence: NSGQNLEEDM GQSEQKADPP ATEKTLLEEK VKLEEQLKET VEKYKRALAD Sequence homology: Guinea Pig: 75%; Human: 100%.
GRPEL1 (GrpE Like 1, Mitochondrial) is a Protein Coding gene. Diseases associated with GRPEL1 include Human Monocytic Ehrlichiosis and Lymphangitis. Among its related pathways are Metabolism of proteins and Mitochondrial protein import. Gene Ontology (GO) annotations related to this gene include protein homodimerization activity and chaperone binding. An important paralog of this gene is GRPEL2.
Specifications
| GRPEL1 | |
| Polyclonal | |
| Unconjugated | |
| GRPEL1 | |
| AA408748; FLJ25609; GREPEL1; GrpE; GrpE like 1, mitochondrial; grpE protein homolog 1, mitochondrial; GrpE#1; GrpE1; GRPEL1; GrpE-like 1; GrpE-like 1, mitochondrial; GrpE-like protein cochaperone; HMGE; mt-GrpE#1; mt-Grpel1; stress-inducible chaperone mt-GrpE#1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 80273 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9HAV7 | |
| GRPEL1 | |
| Synthetic peptide directed towards the N-terminal of human GRPEL1 (aa 35-84). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction