missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GRP94 Recombinant Rabbit Monoclonal Antibody (9U3N5)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA557944

 View more versions of this product

Product Code. 30283700

  • £363.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: EPLLNWMKDK ALKDKIEKAV VSQRLTESPC ALVASQYGWS GNMERIMKAQ AYQTGKDIST NYYASQKKTF EINPRHPLIR DMLRRIKEDE DDKTVLDLAV.

GRP94 is a 803 amino acid protein belonging to the heat shock protein 90 family. It acts as a molecular chaperone that functions in the processing and transport of secreted proteins. GRP94 and its N-terminal fragment stimulates CTL expansion and maturation of human monocyte-derived dendritic cells (MDDC). It plays a central role in innate as well as acquired immunity, maturation and chemotaxis of dendritic cells, Ab production, cross-priming, as a potential marker in breast cancer and is a peptide acceptor in endoplasmic reticulum and an accessory to peptide loading of MHC class I molecules. Expression of GRP94 suppressed A23187-induced apoptosis and stabilized calcium homeostasis. GRP94 is expressed in melanoma or liver metastases of colon carcinoma cells, human gastric carcinoma BGC-823 cells.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GRP94
Recombinant Monoclonal
1.57 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
P08113, P14625, Q66HD0
HSP90B1
A synthetic peptide corresponding to a sequence within amino acids 616-715 of human Grp94 (NP_003290.1).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
ELISA, Western Blot
9U3N5
Unconjugated
HSP90B1
108K heat shock protein; 108K HSP; 94 kDa glucose-regulated protein; 94 kDa glucose-regulated protein {ECO:0000250; 98 kDa protein kinase; ecgp; endoplasmic reticulum resident protein 99; Endoplasmin; endoplasmin {ECO:0000250; endoplasmin-like protein; endothelial cell (HBMEC) glycoprotein; epididymis luminal protein 35; epididymis secretory sperm binding protein Li 125m; ERp99; glucose-regulated protein; glucose-regulated protein GRP94; glucose-regulated protein GRP94 precursor; GP96; gp96 homolog; gp96/GRP94; GRP94; GRP-94; heat shock 108 kDa protein; heat shock protein 108; heat shock protein 108.; heat shock protein 90 beta family member 1; heat shock protein 90 kDa beta member 1; heat shock protein 90 kDa beta member 1 {ECO:0000250; heat shock protein 90, beta (Grp94), member 1; heat shock protein 90, beta, member 1; heat shock protein 90kDa beta (Grp94), member 1; heat shock protein 90kDa beta family member 1; heat shock protein 90kDa beta family member 1 L homeolog; heat shock protein 90kDa beta, member 1; heat shock protein gp96; HEL35; HEL-S-125m; HSP 108; hsp108; HSP90 beta; hsp90b1; hsp90b1 {ECO:0000250; hsp90b1.L; Polymorphic tumor rejection antigen 1; PPK 98; Ppk 98; a protein kinase; ppk98; stress-inducible tumor rejection antigen gp96; TA-3; Targ2; tra1; Tra-1; transferrin-binding protein; transforming growth factor alpha regulated gene 2; tumor rejection antigen; tumor rejection antigen (gp96) 1; tumor rejection antigen 1; Tumor rejection antigen gp96; UniProtKB:P08113}; XELAEV_18017384mg
Rabbit
Protein A
RUO
22027, 362862, 7184
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.