missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GRP94 Recombinant Rabbit Monoclonal Antibody (7P4V7)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557946
This item is not returnable.
View return policy
Description
Immunogen sequence: EPLLNWMKDK ALKDKIEKAV VSQRLTESPC ALVASQYGWS GNMERIMKAQ AYQTGKDIST NYYASQKKTF EINPRHPLIR DMLRRIKEDE DDKTVLDLAV.
GRP94 is a 803 amino acid protein belonging to the heat shock protein 90 family. It acts as a molecular chaperone that functions in the processing and transport of secreted proteins. GRP94 and its N-terminal fragment stimulates CTL expansion and maturation of human monocyte-derived dendritic cells (MDDC). It plays a central role in innate as well as acquired immunity, maturation and chemotaxis of dendritic cells, Ab production, cross-priming, as a potential marker in breast cancer and is a peptide acceptor in endoplasmic reticulum and an accessory to peptide loading of MHC class I molecules. Expression of GRP94 suppressed A23187-induced apoptosis and stabilized calcium homeostasis. GRP94 is expressed in melanoma or liver metastases of colon carcinoma cells, human gastric carcinoma BGC-823 cells.
Specifications
| GRP94 | |
| Recombinant Monoclonal | |
| 1.57 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P08113, P14625, Q66HD0 | |
| HSP90B1 | |
| A synthetic peptide corresponding to a sequence within amino acids 616-715 of human HSP90B1 (NP_003290.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 7P4V7 | |
| Unconjugated | |
| HSP90B1 | |
| 108K heat shock protein; 108K HSP; 94 kDa glucose-regulated protein; 94 kDa glucose-regulated protein {ECO:0000250; 98 kDa protein kinase; ecgp; endoplasmic reticulum resident protein 99; Endoplasmin; endoplasmin {ECO:0000250; endoplasmin-like protein; endothelial cell (HBMEC) glycoprotein; epididymis luminal protein 35; epididymis secretory sperm binding protein Li 125m; ERp99; glucose-regulated protein; glucose-regulated protein GRP94; glucose-regulated protein GRP94 precursor; GP96; gp96 homolog; gp96/GRP94; GRP94; GRP-94; heat shock 108 kDa protein; heat shock protein 108; heat shock protein 108.; heat shock protein 90 beta family member 1; heat shock protein 90 kDa beta member 1; heat shock protein 90 kDa beta member 1 {ECO:0000250; heat shock protein 90, beta (Grp94), member 1; heat shock protein 90, beta, member 1; heat shock protein 90kDa beta (Grp94), member 1; heat shock protein 90kDa beta family member 1; heat shock protein 90kDa beta family member 1 L homeolog; heat shock protein 90kDa beta, member 1; heat shock protein gp96; HEL35; HEL-S-125m; HSP 108; hsp108; HSP90 beta; hsp90b1; hsp90b1 {ECO:0000250; hsp90b1.L; Polymorphic tumor rejection antigen 1; PPK 98; Ppk 98; a protein kinase; ppk98; stress-inducible tumor rejection antigen gp96; TA-3; Targ2; tra1; Tra-1; transferrin-binding protein; transforming growth factor alpha regulated gene 2; tumor rejection antigen; tumor rejection antigen (gp96) 1; tumor rejection antigen 1; Tumor rejection antigen gp96; UniProtKB:P08113}; XELAEV_18017384mg | |
| Rabbit | |
| Protein A | |
| RUO | |
| 22027, 362862, 7184 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction