Learn More
GRP94 Mouse anti-Human, Unconjugated, Clone: 4G7C7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60012-2-IG-150UL
This item is not returnable.
View return policy
Description
The GRP94 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GRP94. This antibody recognizes human antigen. The GRP94 antibody has been validated for the following applications: FC, IF, IHC, WB, ELISA analysis.
Specifications
| GRP94 | |
| Monoclonal | |
| 853 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry | |
| 4G7C7 | |
| Unconjugated | |
| 150 μL | |
| 7184 | |
| IgG1 |