missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GRP78 Recombinant Rabbit Monoclonal Antibody (JE01-35)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA556545

Product Code. 30276300

  • £434.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

GRP78 Recombinant Monoclonal Antibody for Western Blot, IHC (P)

GRP78 is a 78 kDa glucose regulated protein that belongs to the family of heat shock proteins that include HSP70. GRP78 is a resident protein of the endoplasmic reticulum (ER) and associates transiently with a variety of newly synthesized secretory and membrane proteins. GRP78 may also interact with mutant and misfolded proteins, marking them for degradation. The highly conserved sequence Lys-Asp-Glu-Leu (KDEL) is present at the C-terminus of GRP78 and other resident ER proteins including glucose regulated protein 94 (GRP94) and protein disulfide isomerase (PDI). The KDEL signal is recognized by the KDEL receptor and this interaction ensures recycling of escaped ER proteins back to the ER. GRP78's involvement in correct folding of proteins and degradation of misfolded proteins is via its interaction with DNAJC10, probably to facilitate the release of DNAJC10 from its substrate. GRP78 is critical for maintenance of cell homeostasis and the prevention of apoptosis. GRP78 protein levels have been shown to be a reliable biomarker of hypoglycemia and serves a neuroprotective function in neurons exposed to glutamate and oxidative stress. GRP78 levels are reduced in the brains of Alzheimer's Disease patients, and decreased expression of GRP78 is found associated with missense mutations in the human presenilin-1 (PS1) gene. Further, the induction of the GRP78 protein has been associated with the development of drug-resistance to antitumor drugs.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GRP78
Recombinant Monoclonal
1 mg/mL
TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
P06761, P11021, P20029
Hspa5
Recombinant protein within human grp78/bip aa 1-100/654.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Immunohistochemistry (Paraffin), Western Blot
JE01-35
Unconjugated
Hspa5
78 kDa glucose-regulated protein; 78 kDa glucose-regulated protein homolog; 78-kD glucose-regulated protein precursor; AL022860; AU019543; baffled; Binding-immunoglobulin protein; bip; BiP protein; BiP/Grp78; cb865; D2Wsu141e; D2Wsu17e; Endoplasmic reticulum chaperone BiP; endoplasmic reticulum chaperone BiP; 78 kDa glucose-regulated protein; endoplasmic reticulum lumenal Ca(2+)-binding protein grp78; epididymis secretory sperm binding protein Li 89n; fb60h09; fi36d04; FLJ26106; glucose regulated protein, 78 kDa; glucose-regulated protein; glucose-regulated protein, 78kDa; GRP 78; grp78; GRP-78; heat shock 70 kDa protein 5; heat shock 70 kDa protein 5a; heat shock 70kD protein 5; heat shock 70kD protein 5 (glucose-regulated protein, 78kD); heat shock 70kDa protein 5; heat shock 70kDa protein 5 (glucose-regulated protein); heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa); heat shock protein 5; heat shock protein 70 family protein 5; heat shock protein family A (Hsp70) member 5; heat shock protein family A (Hsp70) member 5 L homeolog; heat shock protein family A member 5; heavy-chain binding protein BiP; HEL-S-89n; Hsce70; hsp70; Hsp70 family ATPase KAR2; HSP70 family protein 5; HSPA 5; HSPA5; hspa5.L; hspa5a; I79_019946; immunoglobulin binding protein; immunoglobulin heavy chain-binding protein; Immunoglobulin heavy chain-binding protein homolog; J1248; KAR2; mBiP; MIF2; Sez7; SEZ-7; SSD1; Steroidogenesis-activator polypeptide; wu:fb60h09; wu:fi36d04; XAP-1; XAP-1 antigen; XELAEV_180384511mg; YJL034W; zgc:55994; zgc:77606
Rabbit
Protein A
RUO
14828, 25617, 3309
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.