missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRP78/HSPA5 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 25 publications
£205.00 - £497.00
Specifications
| Antigen | GRP78/HSPA5 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 0.5 ug/ml, Simple Western 1:25, Flow Cytometry 1:150. Use reported in scientific literature (PMID 20208072), Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:50. Use reported in scientific literature (PMID 24089213), Immunohistochemistry-Paraffin reported in scientific literature (PMID 24089213), Immunohistochemistry Free-Floating reported in scientific literature (PMID 26329458) |
| Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Free Floating) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18019123
|
Novus Biologicals
NBP1-06274SS |
0.025 mL |
£205.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18708453
|
Novus Biologicals
NBP1-06274 |
0.1 mL |
£497.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GRP78/HSPA5 Polyclonal specifically detects GRP78/HSPA5 in Human, Mouse, Rat, C. elegans, Chicken, Drosophila, Sheep, Zebrafish samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry - Free Floating.Specifications
| GRP78/HSPA5 | |
| Western Blot 0.5 ug/ml, Simple Western 1:25, Flow Cytometry 1:150. Use reported in scientific literature (PMID 20208072), Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:50. Use reported in scientific literature (PMID 24089213), Immunohistochemistry-Paraffin reported in scientific literature (PMID 24089213), Immunohistochemistry Free-Floating reported in scientific literature (PMID 26329458) | |
| Polyclonal | |
| Rabbit | |
| Cancer, Cellular Markers, ER Markers, Lipid and Metabolism, Unfolded Protein Response, Unfolded Protein Response (UPR) | |
| PBS with 0.02% Sodium Azide | |
| 78 kDa glucose-regulated protein, BiP, Endoplasmic reticulum lumenal Ca(2+)-binding protein grp78, FLJ26106, GRP-78, GRP78BIP, Heat shock 70 kDa protein 5, heat shock 70kD protein 5 (glucose-regulated protein, 78kD), heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa), Immunoglobulin heavy chain-binding protein, MIF2 | |
| HSPA5 | |
| IgG | |
| Affinity Purified | |
| Human, mouse, rat, sheep and chicken. Predicted to react with Zebrafish, S. cerevisiae, primate, bovine, and Drosophila based on 100% sequence homology. |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Free Floating) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, C. elegans, Chicken, Drosophila, Sheep | |
| P11021 | |
| 3309 | |
| Synthetic peptide made to an internal portion of human GRP78 (within residues 250-300). [Swiss-Prot# P11021] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 78 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title