missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRP78/HSPA5 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 7 publications
£184.00 - £427.00
Specifications
| Antigen | GRP78/HSPA5 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 2 ug/ml, Simple Western 1:25, Immunocytochemistry/Immunofluorescence 1:20, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18053704
|
Novus Biologicals
NB300-520SS |
0.025 mL |
£184.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18728773
|
Novus Biologicals
NB300-520 |
0.1 mL |
£427.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GRP78/HSPA5 Polyclonal specifically detects GRP78/HSPA5 in Human, Mouse, Rat, Primate (Negative) samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| GRP78/HSPA5 | |
| Western Blot 2 ug/ml, Simple Western 1:25, Immunocytochemistry/Immunofluorescence 1:20, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Rabbit | |
| Cancer, Cellular Markers, ER Markers, Lipid and Metabolism, Unfolded Protein Response | |
| PBS and 30% Glycerol with 0.05% Sodium Azide | |
| 3309 | |
| A synthetic peptide made to a C-terminal portion of rat GRP78 (within residues 600-654). [Uniprot: P06761] | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| 78 kDa |
| 1 mg/mL | |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Primate (Negative) | |
| 78 kDa glucose-regulated protein, BiP, Endoplasmic reticulum lumenal Ca(2+)-binding protein grp78, FLJ26106, GRP-78, GRP78BIP, Heat shock 70 kDa protein 5, heat shock 70kD protein 5 (glucose-regulated protein, 78kD), heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa), Immunoglobulin heavy chain-binding protein, MIF2 | |
| HSPA5 | |
| IgG | |
| Affinity Purified | |
| Mouse and rat. Does not cross-react with Human or African Green Monkey (COS cells). |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title