missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRP78/HSPA5 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP1-06277SS
This item is not returnable.
View return policy
Description
GRP78/HSPA5 Polyclonal specifically detects GRP78/HSPA5 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunocytochemistry/Immunofluorescence.
Specifications
| GRP78/HSPA5 | |
| Polyclonal | |
| Unconjugated | |
| PBS and 30% Glycerol with 0.1% Sodium Azide | |
| HSPA5 | |
| Synthetic peptide made to a C-terminal portion of rat GRP78 (within residues 600-654). [Swiss-Prot# P06761] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Mouse and rat. Reacts weakly with human. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 1.0 mg/ml | |
| Western Blot 0.5 ug/ml, Simple Western 1:25, Immunocytochemistry/Immunofluorescence 1:50 | |
| 78 kDa glucose-regulated protein, BiP, Endoplasmic reticulum lumenal Ca(2+)-binding protein grp78, FLJ26106, GRP-78, GRP78BIP, Heat shock 70 kDa protein 5, heat shock 70kD protein 5 (glucose-regulated protein, 78kD), heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa), Immunoglobulin heavy chain-binding protein, MIF2 | |
| Rabbit | |
| 78 kDa | |
| 0.025 mL | |
| Cancer, Cellular Markers, ER Markers, Lipid and Metabolism, Unfolded Protein Response | |
| 3309 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction