missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRP78/BIP Mouse anti-Human, Rat, Mouse, CoraLite 650, Clone: 1D6F7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL650-66574-100UL
This item is not returnable.
View return policy
Description
GRP78/BIP Monoclonal antibody apecifically detetects GRP78/BIP antigen in Human, Rat, Mouse Samples. It is validated for Flow Cytometry.Specifications
| GRP78/BIP | |
| Monoclonal | |
| 1000 μg/mL | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| HSPA5 | |
| GRP78/BIP fusion protein Ag2188 | |
| 100 μL | |
| 3309 | |
| Store at -20°C. Avoid exposure to light. | |
| IgG1 |
| Flow Cytometry | |
| 1D6F7 | |
| CoraLite 650 | |
| BC020235 | |
| Mouse | |
| Protein G purification | |
| Primary | |
| Human, Rat, Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction