missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRP75/HSPA9B/Mortalin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-31596
This item is not returnable.
View return policy
Description
GRP75/HSPA9B/Mortalin Polyclonal specifically detects GRP75/HSPA9B/Mortalin in Human, Mouse, Rat, Porcine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| GRP75/HSPA9B/Mortalin | |
| Polyclonal | |
| Unconjugated | |
| 1XPBS, 1% BSA, 10% Glycerol (pH7). with 0.025% Proclin 300 | |
| HSPA9 | |
| Recombinant protein encompassing a sequence within the center region of human GRP75. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Tested in human. Predicted cross-reactivity based on sequence identity: Zebrafish (87%) and Xenopus tropicalis (94%). Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 0.19mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation 1:100-1:500, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| 75 kDa glucose-regulated protein, GRP-75, GRP75mthsp75, Heat shock 70 kDa protein 9, heat shock 70kD protein 9B, heat shock 70kDa protein 9 (mortalin), heat shock 70kDa protein 9B (mortalin-2), HSPA9BMGC4500, Mortalin, mortalin, perinuclear, mortalin-2, MOT, mot-2, MOT2, MTHSP75, p66-mortalin, PBP74CSA, Peptide-binding protein 74, stress-70 protein, mitochondrial | |
| Rabbit | |
| 74 kDa | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 3313 | |
| Human, Mouse, Pig | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction