missing translation for 'onlineSavingsMsg'
Learn More

GRP75/HSPA9B/Mortalin Antibody, Novus Biologicals™

Product Code. 18402451 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18402451 25 μL 25µL
18206166 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18402451 Supplier Novus Biologicals Supplier No. NBP18972925ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 1 publication

GRP75/HSPA9B/Mortalin Polyclonal specifically detects GRP75/HSPA9B/Mortalin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRP75/HSPA9B/Mortalin
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias 75 kDa glucose-regulated protein, GRP-75, GRP75mthsp75, Heat shock 70 kDa protein 9, heat shock 70kD protein 9B, heat shock 70kDa protein 9 (mortalin), heat shock 70kDa protein 9B (mortalin-2), HSPA9BMGC4500, Mortalin, mortalin, perinuclear, mortalin-2, MOT, mot-2, MOT2, MTHSP75, p66-mortalin, PBP74CSA, Peptide-binding protein 74, stress-70 protein, mitochondrial
Gene Symbols HSPA9
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:GIVHVSAKDKGTGREQQIVIQSSGGLSKDDIENMVKNAEKYAEEDRRKKERVEAVNMAEGIIHDTETKMEEFKDQLPADECNKLKEEISKMRELLARKDSETGENIRQAASSLQQASLKLFEMAYKKMASEREGSGSSGT
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3313
Test Specificity Specificity of human GRP75/HSPA9B/Mortalin antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.