missing translation for 'onlineSavingsMsg'
Learn More

GRP75/HSPA9B/Mortalin Antibody (419612) [Janelia Fluor™ 646], Novus Biologicals™

Product Code. 30733257 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30733257 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733257 Supplier Novus Biologicals Supplier No. FAB3584J

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GRP75/HSPA9B/Mortalin Monoclonal antibody specifically detects GRP75/HSPA9B/Mortalin in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRP75/HSPA9B/Mortalin
Applications Western Blot
Classification Monoclonal
Clone 419612
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias 75 kDa glucose-regulated protein, GRP-75, GRP75mthsp75, Heat shock 70 kDa protein 9, heat shock 70kD protein 9B, heat shock 70kDa protein 9 (mortalin), heat shock 70kDa protein 9B (mortalin-2), HSPA9BMGC4500, Mortalin, mortalin, perinuclear, mortalin-2, MOT, mot-2, MOT2, MTHSP75, p66-mortalin, PBP74CSA, Peptide-binding protein 74, stress-70 protein, mitochondrial
Host Species Mouse
Immunogen E. coli-derived recombinant human GRP75/HSPA9B, Glu542-Gln679, Accession # P38646
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3313
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.