missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRP75/HSPA9B/Mortalin Antibody (30A5), Novus Biologicals™
Mouse Monoclonal Antibody
£135.00 - £724.00
Specifications
| Antigen | GRP75/HSPA9B/Mortalin |
|---|---|
| Clone | 30A5 |
| Dilution | Western Blot 1:1000, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin 1:10-1:500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18095879
|
Novus Biologicals
NBP1-97448-0.025ML |
0.025 mL |
£135.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18120990
|
Novus Biologicals
NBP1-97448-0.05ML |
0.05 mL |
£409.00
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18087709
|
Novus Biologicals
NBP1-97448-0.2ML |
0.2 mL |
£724.00
0.20mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GRP75/HSPA9B/Mortalin Monoclonal specifically detects GRP75/HSPA9B/Mortalin in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Drosophila, Guinea Pig, Hamster, Primate, Rabbit, Sheep, Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| GRP75/HSPA9B/Mortalin | |
| Western Blot 1:1000, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Hamster, Pig, Human, Chicken, Guinea Pig, Rat, Sheep, Primate, Bovine, Rabbit, Xenopus species, Canine, Drosophila, Mouse | |
| 75 kDa glucose-regulated protein, GRP-75, GRP75mthsp75, Heat shock 70 kDa protein 9, heat shock 70kD protein 9B, heat shock 70kDa protein 9 (mortalin), heat shock 70kDa protein 9B (mortalin-2), HSPA9BMGC4500, Mortalin, mortalin, perinuclear, mortalin-2, MOT, mot-2, MOT2, MTHSP75, p66-mortalin, PBP74CSA, Peptide-binding protein 74, stress-70 protein, mitochondrial | |
| HSPA9 | |
| IgG1 | |
| Ammonium sulfate precipitation |
| 30A5 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Core ESC Like Genes, Stem Cell Markers | |
| PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide | |
| 3313 | |
| Native human Grp75 (Mortalin). | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title