missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Growth Hormone Releasing Factor (GRF) (1-29) amide, human

Product Code. 15400177
Click to view available options
Quantity:
0.5 mg
1 mg
Unit Size:
0.5mg
1mg
This item is not returnable. View return policy

Product Code. 15400177

Brand: Thermo Scientific Chemicals J61111.!D

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A peptide hormone that stimulates release of the growth hormone

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86168-78-7

Specifications

Content And Storage Ambient temperatures
Format Powder
Molecular Formula C151H250N44O44S
MDL Number MFCD00076559
InChI Key BVLCEKWPOSAKSZ-YQMCHIOTSA-N
SMILES CC(O)=O.CC[C@H](C)[C@H](NC(=O)[C@H](C)NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](C)NC(=O)[C@@H](N)CC1=CC=C(O)C=C1)C(=O)N[C@@H](CC1=CC=CC=C1)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](CO)C(=O)N[C@@H](CC1=CC=C(O)C=C1)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CC(C)C)C(=O)NCC(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CO)C(=O)N[C@@H](C)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H]([C@@H](C)CC)C(=O)N[C@@H](CCSC)C(=O)N[C@@H](CO)C(=O)N[C@@H](CCCNC(N)=N)C(N)=O
IUPAC Name (3S)-3-{[(1S)-1-{[(1S,2S)-1-{[(1S)-1-{[(1S,2R)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-5-amino-1-{[(1S)-1-{[(1S)-1-[({[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-5-amino-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S,2S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-4-carbamimidamido-1-carbamoylbutyl]carbamoyl}-2-hydroxyethyl]carbamoyl}-3-(methylsulfanyl)propyl]carbamoyl}-2-methylbutyl]carbamoyl}-2-carboxyethyl]carbamoyl}-3-carbamoylpropyl]carbamoyl}-3-methylbutyl]carbamoyl}-3-methylbutyl]carbamoyl}pentyl]carbamoyl}-4-carbamimidamidobutyl]carbamoyl}ethyl]carbamoyl}-2-hydroxyethyl]carbamoyl}-3-methylbutyl]carbamoyl}-3-carbamoylpropyl]carbamoyl}methyl)carbamoyl]-3-methylbutyl]carbamoyl}-2-methylpropyl]carbamoyl}pentyl]carbamoyl}-4-carbamimidamidobutyl]carbamoyl}-2-(4-hydroxyphenyl)ethyl]carbamoyl}-2-hydroxyethyl]carbamoyl}-2-carbamoylethyl]carbamoyl}-2-hydroxypropyl]carbamoyl}-2-phenylethyl]carbamoyl}-2-methylbutyl]carbamoyl}ethyl]carbamoyl}-3-[(2S)-2-[(2S)-2-amino-3-(4-hydroxyphenyl)propanamido]propanamido]propanoic acid; acetic acid
Quantity 0.5 mg
Molecular Weight (g/mol) 3417.99

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.