missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Growth hormone Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55243-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody has no cross reaction to CSH1.
Human growth hormone (hGH) is a member of the somatotropin/prolactin family of hormones which play an important role in growth control. The GH1 gene, along with 4 additional related genes, is located at the growth hormone locus on chromosome 17. The five genes share a remarkably high degree of sequence identity. Alternative splicing results in 5 isoforms ranging from approximately 19 kDa to 24 kDa. HGH is produced by the somatotroph cells in the pituitary. It acts through binding to membrane receptors that belong to the cytokine receptor subfamily. Its major role in stimulating body growth is to stimulate the liver and other tissues to secrete IGF-1. It stimulates both the differentiation and proliferation of myoblasts. It also stimulates amino acid uptake and protein synthesis in muscle and other tissues. Mutations or deletions in the gene lead to growth hormone deficiency and short stature.Specifications
| Growth hormone | |
| Polyclonal | |
| Unconjugated | |
| GH1 | |
| GH, GH N, GH1, GHN, Growth hormone, growth hormone 1, hGH N, Pituitary growth hormone, Somatotropin | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2688 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.43 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P01241 | |
| GH1 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction