missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Growth Hormone Mouse, Clone: GH/1450, enQuireBio™
Mouse Monoclonal Antibody
£181.83 - £395.00
Specifications
Antigen | Growth Hormone |
---|---|
Clone | GH/1450 |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15970088
|
EnquireBio
2688-MSM3-P1 |
100 μg |
£395.00
100µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15960088
|
EnquireBio
2688-MSM3-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Growth Hormone | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
2688 | |
A recombinant human Growth Hormone (GH) fragment (aa58-187) | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
20kDa |
GH/1450 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
NP_072053 | |
GH1 | |
IgG2b | |
Protein A/G Purified | |
Cytoplasmic | |
Pituitary cells or Pituitary |