missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Growth Hormone ELISA Kit

Product Code. 15475003
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
1 set
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15475003 96 Tests 1 set
17105459 5 x 96 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15475003 Supplier Invitrogen™ Supplier No. EHGH1
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ELISA

The Human Growth Hormone (Hu GH) ELISA quantitates Hu GH in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu GH. Principle of the method The Human GH solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Human growth hormone (hGH) is a member of the somatotropin/prolactin family of hormones which play an important role in growth control. The GH1 gene, along with 4 additional related genes, is located at the growth hormone locus on chromosome 17. The five genes share a remarkably high degree of sequence identity. Alternative splicing results in 5 isoforms ranging from approximately 19 kDa to 24 kDa. HGH is produced by the somatotroph cells in the pituitary. It acts through binding to membrane receptors that belong to the cytokine receptor subfamily. Its major role in stimulating body growth is to stimulate the liver and other tissues to secrete IGF-1. It stimulates both the differentiation and proliferation of myoblasts. It also stimulates amino acid uptake and protein synthesis in muscle and other tissues. Mutations or deletions in the gene lead to growth hormone deficiency and short stature.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01241
Assay Range 4-600 pg/mL
Assay Sensitivity 4 pg/mL
Calibration Range 4 pg/mL to 600 pg/mL
Content And Storage Pre-coated plate(s), standard or calibrator, detector antibody, HRP conjugate, diluents, wash buffer, substrate(s), and stop solution. Store kit at 2-8°C. See product manual for detailed contents and storage conditions for maximum stability.
Assay Solid-phase sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Growth Factors & Receptors
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA kit shows no cross-reactivity with any of the cytokines tested: Human Angiogenin, BDNF, BLC, ENA-78, FGF-4, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-7, IL-8, IL-9, IL-10, IL-11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, G-CSF, GM-CSF, IFN-gamma, Leptin, MCP-1, MCP-2, MCP-3, MDC, MIP-1 alpha, MIP-1 beta, MIP-1 delta, PARC, PDGF, RANTES, SCF, TARC, TGF-beta, TIMP-1, TIMP-2, TNF-alpha, TNF-beta, TPO, VEGF.
Sufficient For 96 assays
Target Species Human
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias dog growth hormone; GH; Gh1; GHB1; GHB5; GHN; GH-N; Growth hormone; growth hormone 1; growth hormone 12; growth hormone B1; growth hormone B5; growth hormone precursor; HGH; hGH-N; IGHD1B; pituitary growth hormone; rHGH; RNGHGP; Somatotropindog growth hormone; GH; Gh1; GHB1; GHB5; GHN; GH-N; Growth hormone; growth hormone 1; growth hormone 12; growth hormone B1; growth hormone B5; growth hormone precursor; HGH; hGH-N; IGHD1B; pituitary growth hormone; rHGH; RNGHGP; Somatotropin; GH; GH-N; GHB5; GHN; IGHD1B; hGH-N
Gene ID (Entrez) 2688
Gene Symbol Gh1
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Protein Aliases Growth hormone, GH, GH-N, Growth hormone 1, Pituitary growth hormone
Quantity 96 Tests
Regulatory Status RUO
Research Discipline Biomarker-Related Pathways
Sample Volume Plasma 25 μL, Serum 25 μL, Supernatant 100 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Synonym GH, GH-N, GHN, IGHD1B, hGH-N
Target Growth Hormone
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.